Skip to Content
Merck
All Photos(3)

Key Documents

ALD00176

Sigma-Aldrich

4-(Acetylamino)benzenesulfonyl fluoride

95%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€956.00

€956.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€956.00

About This Item

Empirical Formula (Hill Notation):
C8H8FNO3S
CAS Number:
Molecular Weight:
217.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€956.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

173-178 °C

SMILES string

FS(C1=CC=C(NC(C)=O)C=C1)(=O)=O

InChI

1S/C8H8FNO3S/c1-6(11)10-7-2-4-8(5-3-7)14(9,12)13/h2-5H,1H3,(H,10,11)

InChI key

HRQHLUBMODFETM-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article details the latest progress in synthesizing alkylsulfonyl fluorides through different approaches that utilize photoredox catalysis, electrocatalysis, transition-metal catalysis, and organocatalysis. The alkylsulfonyl fluorides thus prepared could be utilized further in sulfur(VI) fluoride exchange (SuFEx) click reactions.

In collaboration with K. Barry Sharpless and coworkers, we offer a variety of sulfonyl fluoride reagents that undergo a “Click II” reaction.

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service