Skip to Content
Merck
All Photos(1)

Documents

ALD00140

Sigma-Aldrich

3,4-Dimethoxybenzenesulfonyl fluoride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9FO4S
CAS Number:
Molecular Weight:
220.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

59-63 °C

SMILES string

FS(C1=CC(OC)=C(OC)C=C1)(=O)=O

InChI

1S/C8H9FO4S/c1-12-7-4-3-6(14(9,10)11)5-8(7)13-2/h3-5H,1-2H3

InChI key

LCEVVNNKUAEPJM-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service