Skip to Content
Merck
All Photos(2)

Key Documents

ALD00098

Sigma-Aldrich

3,6-Bis(methoxycarbonyl)-1,2,4,5-tetrazine

Synonym(s):

Dimethyl 1,2,4,5-tetrazine-3,6-dicarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€754.00

€754.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€754.00

About This Item

Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€754.00


Check Cart for Availability

Request a Bulk Order

form

crystals

storage temp.

−20°C

SMILES string

O=C(C1=NN=C(C(OC)=O)N=N1)OC

InChI

1S/C6H6N4O4/c1-13-5(11)3-7-9-4(10-8-3)6(12)14-2/h1-2H3

InChI key

QODPSGGFQFBWME-UHFFFAOYSA-N

Application

Tetrazine has shown to be a versatile building block as a substrate in inverse elecron demand hetero Diels-Alder reactions. The Boger group has reported this powerful approach in the synthesis of alkaloids.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Boger Lab utilizes nature's solutions in complex natural products, extending them through rational design for improved biological outcomes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service