Skip to Content
Merck
All Photos(1)

Key Documents

ALD00012

Sigma-Aldrich

Benzyl (E)-2-(2-oxobenzofuran-3(2H)-ylidene)acetate

95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€8.70

€8.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€8.70

About This Item

Empirical Formula (Hill Notation):
C17H12O4
CAS Number:
Molecular Weight:
280.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€8.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95% (HPLC)

form

flakes

storage temp.

2-8°C

SMILES string

O=C(OCC1=CC=CC=C1)/C=C(C2=C(O3)C=CC=C2)/C3=O

InChI

1S/C17H12O4/c18-16(20-11-12-6-2-1-3-7-12)10-14-13-8-4-5-9-15(13)21-17(14)19/h1-10H,11H2/b14-10+

InChI key

HXSOJKAKJXISCE-GXDHUFHOSA-N

Application

Building block used as a substrate in phosphine-catalyzed domino sequences. This versatile compound can be used to synthesize a wide scope of spirocyclic benzofuranone scaffolds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service