Skip to Content
Merck
All Photos(2)

Key Documents

A94638

Sigma-Aldrich

1-Aza-2-cyclooctanone

98%

Synonym(s):

2-Azacyclooctanone, 2-Azocanone, 2-Oxo-heptamethyleneimine, Enantholactam, Hexahydro-2(1H)-azocinone, Oenantholactam

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€81.70

€81.70


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
€81.70

About This Item

Empirical Formula (Hill Notation):
C7H13NO
CAS Number:
Molecular Weight:
127.18
Beilstein:
108987
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€81.70


0 left in stock

Request a Bulk Order

Assay

98%

form

crystals

bp

148-150 °C/10 mmHg (lit.)

mp

35-38 °C (lit.)

SMILES string

O=C1CCCCCCN1

InChI

1S/C7H13NO/c9-7-5-3-1-2-4-6-8-7/h1-6H2,(H,8,9)

InChI key

CJYXCQLOZNIMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformational energy studies of the growth hormone inhibitor, cyclo (Aha-Cys-Phe-D-Trp-Lys-Thr-Cys).
F A Momany
Biochemical and biophysical research communications, 95(1), 61-66 (1980-07-16)
Lothar Schwarz et al.
Journal of forensic sciences, 52(3), 649-655 (2007-04-26)
A new method for enhancement of ninhydrin or 1,8-diazafluoren-9-one (DFO)-treated latent fingerprints on thermal paper will be described. Most thermosensitive surfaces of thermal paper become dark when treated with DFO or ninhydrin petroleum ether (NPB) solution. This effect minimizes contrast
Yan Huang et al.
Journal of biomedical science, 16, 87-87 (2009-09-24)
Stem cell therapy has emerged as a potential therapeutic option for tissue engineering and regenerative medicine, but many issues remain to be resolved, such as the amount of seed cells, committed differentiation and the efficiency. Several previous studies have focused

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service