Skip to Content
Merck
All Photos(1)

Key Documents

A89901

Sigma-Aldrich

2-Aminobenzonitrile

98%

Synonym(s):

2-Cyanoaniline, Anthranilonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€51.10
100 G
€144.00

€51.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€51.10
100 G
€144.00

About This Item

Linear Formula:
H2NC6H4CN
CAS Number:
Molecular Weight:
118.14
Beilstein:
907187
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€51.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

bp

267-268 °C (lit.)

mp

45-48 °C (lit.)

SMILES string

Nc1ccccc1C#N

InChI

1S/C7H6N2/c8-5-6-3-1-2-4-7(6)9/h1-4H,9H2

InChI key

HLCPWBZNUKCSBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chrisoula Kakoulidou et al.
Journal of inorganic biochemistry, 211, 111194-111194 (2020-08-01)
The interaction of the novel quinazoline (E)-4-(2-(pyridin-2-ylmethylene)hydrazinyl)quinazoline (L) with Zn2+ was performed in the absence or presence of the non-steroidal anti-inflammatory drug sodium diclofenac (Nadicl) and resulted in the formation of complexes [Zn(L)2](NO3)2·MeOH (1·MeOH) and [Zn(L)(dicl-O)2]·MeOH (2·MeOH), respectively. The two
Jiayin Hu et al.
ChemSusChem, 11(24), 4219-4225 (2018-11-16)
The efficient transformation of CO2 into value-added chemicals with green, abundant, and cheap catalysts is an interesting and challenging topic in both green and sustainable chemistry. In this study, a series of salt-lake brines were used for the first time

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service