A80807
2-Amino-2-thiazoline
97%
Synonym(s):
2-Thiazolin-2-amine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H6N2S
CAS Number:
Molecular Weight:
102.16
Beilstein:
105367
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
mp
79-82 °C (lit.)
SMILES string
NC1=NCCS1
InChI
1S/C3H6N2S/c4-3-5-1-2-6-3/h1-2H2,(H2,4,5)
InChI key
REGFWZVTTFGQOJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A S Sobolev et al.
Radiation and environmental biophysics, 23(2), 79-93 (1984-01-01)
The sulphur-containing radioprotectors mercaptoethylamine (MEA), aminoethylisothiourea (AET), 2- aminothiazoline , 4-oxo-2- aminothiazoline , and S-S-3- oxapentane -1,5- diisothiourea , and the radioprotective biogenic amines serotonin, histamine, and dopamine, caused the elevation of cAMP content and intensified the rate of cAMP-dependent
Zhigang Li et al.
Bioorganic & medicinal chemistry letters, 15(7), 1769-1772 (2005-03-23)
A new family of 2-aminothiazonaphthalimides with different side chains as novel intercalative and visible light activatable photonucleases, was designed, synthesized and quantitatively evaluated. The order of their photocleaving abilities was parallel to that of their intercalative properties. The compound with
L T Zolotareva et al.
Radiobiologiia, 27(2), 189-194 (1987-03-01)
The influence of 2-amino-2-thiazoline on lipid peroxidation in yeasts Saccharomyces cerevisiae and Pichia guilliermondii has been studied in vivo and in vitro. In the case the radioresistance of diploid yeasts-saccharomycetes is changed the radioprotector can produce a direct effect on
L T Zolotareva et al.
Radiobiologiia, 24(5), 654-656 (1984-09-01)
A study was made of the dependence of different natural and modified radioresistance upon glutathione content of yeast cells (Saccharomyces cerevisiae and Pichia guilliermondii). It was shown that glutathione was only involved in the formation of natural radioresistance in Saccharomyces
J J Díaz Gil et al.
Revista espanola de oncologia, 29(4), 615-621 (1982-01-01)
Some aspects of the effect of thioproline and 2-amino-thiazoline-HCl on HeLa cell cultures are studied. Both drugs, at 1 mM concentrations, produce morphological changes that become stabilized after four days. The morphology changes depend on the microtubules, microfilaments, and de
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service