Skip to Content
Merck
All Photos(3)

Key Documents

A59557

Sigma-Aldrich

5-Aminoindazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€54.30

€54.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€54.30

About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€54.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

175-178 °C (lit.)

SMILES string

Nc1ccc2[nH]ncc2c1

InChI

1S/C7H7N3/c8-6-1-2-7-5(3-6)4-9-10-7/h1-4H,8H2,(H,9,10)

InChI key

XBTOSRUBOXQWBO-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chai Hoon Soh et al.
Journal of combinatorial chemistry, 8(4), 464-468 (2006-07-11)
A microwave-assisted parallel synthesis of 2,4-disubstituted 5-aminoimidazoles has been developed. Significant rate enhancement was observed for all steps in the three-step protocol. The overall reaction time was shortened to 25 min, as compared to 53 h for the conventional procedures.
Feng Shi et al.
Molecular diversity, 15(2), 497-505 (2010-09-04)
Six new series of pyrazolo[4,3-f]quinoline derivatives with potential bioactivities were synthesized by the three-component reactions of aromatic aldehydes, 5-aminoindazole, and various cyclic 1,3-dicarbonyl compounds under microwave irradiation. This protocol has the valuable features of structural diversity of products, broader substrate
Ratika Krishnamurty et al.
Bioorganic & medicinal chemistry letters, 21(1), 550-554 (2010-11-17)
Affinity reagents that target protein kinases are powerful tools for signal transduction research. Here, we describe a general set of kinase ligands based on a 5-aminoindazole scaffold. This scaffold can readily be derivatized with diverse binding elements and immobilized analogs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service