Skip to Content
Merck
All Photos(2)

Key Documents

A51807

Sigma-Aldrich

2-Amino-4,6-dimethylpyridine

99%

Synonym(s):

6-Amino-2,4-lutidine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€72.40
25 G
€162.00

€72.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€72.40
25 G
€162.00

About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€72.40


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

99%

form

powder

bp

235 °C (lit.)

mp

63-64 °C (lit.)

SMILES string

Cc1cc(C)nc(N)c1

InChI

1S/C7H10N2/c1-5-3-6(2)9-7(8)4-5/h3-4H,1-2H3,(H2,8,9)

InChI key

BRBUBVKGJRPRRD-UHFFFAOYSA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service