Skip to Content
Merck
All Photos(1)

Key Documents

96835

Sigma-Aldrich

Z-Lys(Z)-OH

≥98.0% (TLC)

Synonym(s):

N,N′-Di-Cbz-L-lysine, Nα,Nε-Di-Z-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2O2CNH(CH2)4CH(NHCO2CH2C6H5)CO2H
CAS Number:
Molecular Weight:
414.45
Beilstein:
1897206
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

[H][C@@](CCCCNC(=O)OCc1ccccc1)(NC(=O)OCc2ccccc2)C(O)=O

InChI

1S/C22H26N2O6/c25-20(26)19(24-22(28)30-16-18-11-5-2-6-12-18)13-7-8-14-23-21(27)29-15-17-9-3-1-4-10-17/h1-6,9-12,19H,7-8,13-16H2,(H,23,27)(H,24,28)(H,25,26)/t19-/m0/s1

InChI key

BLZXFNUZFTZCFD-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service