Skip to Content
Merck
All Photos(1)

Key Documents

96720

Sigma-Aldrich

Z-Tle-OH (dicyclohexylammonium) salt

≥99.0% (HPLC)

Synonym(s):

Z-L-tert-Leucine (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€1,270.00

€1,270.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€1,270.00

About This Item

Linear Formula:
(CH3)3CCH(NHCOOCH2C6H5)COOH · HN(C6H11)2
CAS Number:
Molecular Weight:
446.62
Beilstein:
3831936
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

€1,270.00


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

≥99.0% (HPLC)

form

powder

optical activity

[α]20/D −6.5±1°, c = 1% in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

163-167 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CC(C)(C)[C@H](NC(=O)OCc3ccccc3)C(O)=O

InChI

1S/C14H19NO4.C12H23N/c1-14(2,3)11(12(16)17)15-13(18)19-9-10-7-5-4-6-8-10;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h4-8,11H,9H2,1-3H3,(H,15,18)(H,16,17);11-13H,1-10H2/t11-;/m1./s1

InChI key

ZZVHEFCADXTNTP-RFVHGSKJSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service