Skip to Content
Merck
All Photos(3)

Documents

96092

Sigma-Aldrich

N-Z-1,3-Propanediamine hydrochloride

≥98.0% (AT)

Synonym(s):

N-Z-1,3-Diaminopropane hydrochloride, Benzyl N-(3-aminopropyl)carbamate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCONH(CH2)3NH2 · HCl
CAS Number:
Molecular Weight:
244.72
Beilstein:
4725495
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (AT)

form

crystals

reaction suitability

reagent type: cross-linking reagent

mp

185-189 °C

functional group

Cbz
amine

SMILES string

[H]Cl.NCCCNC(OCC1=CC=CC=C1)=O

InChI

1S/C11H16N2O2.ClH/c12-7-4-8-13-11(14)15-9-10-5-2-1-3-6-10;/h1-3,5-6H,4,7-9,12H2,(H,13,14);1H

InChI key

XKMBTMXQMDLSRB-UHFFFAOYSA-N

Other Notes

Building block

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F R Pfeiffer et al.
Journal of medicinal chemistry, 27(3), 325-341 (1984-03-01)
Tri- and tetrapeptide analogues were synthesized and evaluated as renal vasodilators. These peptides were prepared by standard coupling reactions, which also provided good yields with hindered alpha-methyl amino acid derivatives. Preliminary evidence of renal vasodilator activity was determined in anesthetized
S A Boyd et al.
Journal of medicinal chemistry, 37(19), 2991-3007 (1994-09-16)
The aim of this study was the discovery of nonpeptide renin inhibitors with much improved oral absorption, bioavailability, and efficacy, for use as antihypertensive agents. Our prior efforts led to the identification of A-74273 [1,R = 3-(4-morpholino)propyl], with a bioavailability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service