Skip to Content
Merck
All Photos(1)

Documents

94481

Sigma-Aldrich

1-(2-Methoxyethoxy)-2-methyl-2-propanol

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O3
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

liquid

refractive index

n20/D 1.422

density

0.953 g/mL at 20 °C (lit.)

SMILES string

COCCOCC(C)(C)O

InChI

1S/C7H16O3/c1-7(2,8)6-10-5-4-9-3/h8H,4-6H2,1-3H3

InChI key

SIUWDFVMEASCRP-UHFFFAOYSA-N

Other Notes

Stable precursor for a watersoluble unstable Boc reagent, e.g. the chloroformate

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Birr
Solid Phase Synth. and Combinat. Libr., Coll. Papers, 5th Intl. Synmp., 82-82 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service