Skip to Content
Merck
All Photos(1)

Documents

94154

Sigma-Aldrich

(R)-5-Hexanolide

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(R)-δ-Methyl-δ-valerolactone, (R)-6-Methyltetrahydro-2H-pyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein:
5241702
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D +29.5±2°, c = 1% in chloroform

storage temp.

2-8°C

SMILES string

C[C@@H]1CCCC(=O)O1

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service