Skip to Content
Merck
All Photos(1)

Key Documents

936650

Sigma-Aldrich

Lithium perchlorate

greener alternative

Technipur®, battery grade, dry, 99.99% trace metals basis

Synonym(s):

Lithium cloricum, Lithium salt, Perchloric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiClO4
CAS Number:
Molecular Weight:
106.39
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.21
Pricing and availability is not currently available.

grade

battery grade

Quality Level

Assay

99.99% trace metals basis

form

dry
powder and chunks

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

236 °C (lit.)

solubility

water: 106.4 g/L at 20 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Li/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

MHCFAGZWMAWTNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. Lithium perchlorate (LiClO4) belongs to an Enabling category, and is a sustainable electrolyte salt for lithium-sulfur batteries, inhibiting the polysulfide shuttle and enhancing coulombic efficiency. Its low charge-transfer resistance and protective passivation layer on the lithium anode improve stability and performance, contributing to greener battery technologies. Click here for more information.

Application

Lithium perchlorate (LiClO4) is used as an electrolyte for lithium-sulfur batteries.[1] [2] [3]

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of lithium chlorate salts on structural and electrical properties of natural polymer electrolytes for all solid state lithium polymer batteries.
Perumal P, et al.
Vacuum, 159, 277-281 (2019)
Nonaqueous liquid electrolytes for lithium-based rechargeable batteries
Xu K, et al.
Chemical Reviews, 104(10), 4303-4418 (2004)
Effect of Lithium Perchlorate on the Optoelectrical and Thermal Properties of Poly (Vinylpyrrolidone)/Nano-Cesium Aluminate Solid Polymer Electrolytes
Saeed A M, et al.
Polymer-Plastics Technology and Engineering, 57(15), 1554-1566 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service