Skip to Content
Merck
All Photos(2)

Key Documents

935778

Sigma-Aldrich

Multiphoton lithography bioink kit

polyester-based biodegradable bioink

Synonym(s):

DEGRAD INX X100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

description

Ultimate Strength 5-10 MPa
Youngs modulus 50-60 MPa

Quality Level

color

orange-yellow, liquid

viscosity

0.5 -2 Pa.s

elongation

At break - 20-30%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This polyester-based synthetic resin for multi-photon lithography (MPL) based 3D bioprinting applications combines the benefits of biocompatibility, flexibility, and easy proccessability, resulting in high resolution 3D bioprinted features.

The multiphoton lithography bioink kit is a ready to use formulation that can be processed via a MPL based printer after a short pre-heating process. The resin can be processed at high scanning speeds (up to at least 600mm/s) which is favorable for shorter fabrication times.

Application

Applications:
3D bioprinting
Tissue engineering
High resolution printing of complex and open geometries with mechanical robustness

Features and Benefits

- Biocompatibility - expression biocompatibility with no toxic effect on living cells

- Biodegradability - long term degradation profile when in contact with water, cell culture media, or biological fluids

- Proccessability - Easy processing into open and complex architectures with minimal deformation

- High resolution - Resolution of <500nm

- Flexibility - Can be used to print strong yet flexible structures that are favorable for easy handling and processing

- Easy to handle - Provided as ready to print formulation

- Reproducibility - Production under strict quality control to provide a material that delivers every time

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.9 °F

Flash Point(C)

-14.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier Guillaume et al.
Acta biomaterialia, 165, 72-85 (2022-03-16)
Since its inception, tissue engineering and regenerative medicine (TERM) has been relying on either scaffold-based or scaffold-free strategies. Recent reports outlined the possibility of a synergistic, convergence approach, referred to as the third TERM strategy, which could alleviate bottlenecks of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service