Skip to Content
Merck
All Photos(1)

Key Documents

934801

Sigma-Aldrich

Glycidol chitosan

Degree of functionalization > 80%

Synonym(s):

Dihydroxypropyl chitosan, Glycolchitosan, glycol chitosan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C9H17NO6]n [C8H13NO5]m
UNSPSC Code:
12352201
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

form

powder or chunks (or fibres)

color

white to off-white

storage temp.

2-8°C

General description

Chitosan is a cationic biodgradable and biocompatible biopolymer that has been commonly used in many applications. The polymer chains are positively charged when disolved in aqueous solutions of pH<6.5. However, the biomedical applications of the positively charged chitosan are limited becuase it cannot be easily dissolved in aqueous media. Glycidol chitosan is a derivative of chitosan whereby hydrohilic glycidol is grafted onto the amino groups of chitosan to improve its solubility in a wider range of pH. It possesses many useful biological properties such as biocompatibility, biodegradability, and antimicrobial properties. It can also be further functionalized to induce photo and thermally crosslinkable properites and can be used as a precursor for hydrogel preparations that can be used in many biomedical applications.

Application

  • Hydrogels
  • Tissue engineering
  • Bioprinting
  • Drug delivery system

Features and Benefits

  • Naturally derived biopolymer
  • Cationic
  • Enhanced solubility
  • Tunable precursor material

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chitosan chemistry and pharmaceutical perspectives
Kumar MR
Chemical Reviews, 104 (2004)
Modified chitosan hydrogels as drug delivery and tissue engineering systems: present status and applications
Tapan Kumar Giri
Acta Pharmaceutica Sinica. B, 2 (2012)
Effect of chitosan dissolved in different acids on its ability to control postharvest gray mold of table grape
Romanazzi G
Phytopathology, 99(9) (2009)
Zhilong Shi et al.
Biomaterials, 27(11), 2440-2449 (2005-12-13)
Although total joint replacement has become commonplace in recent years, bacterial infection remains a significant complication following this procedure. One approach to reduce the incidence of joint replacement infection is to add antimicrobial agents to the bone cement used to
Glycol Chitosan: A Water-Soluble Polymer for Cell Imaging and Drug Delivery
Fengming Lin
Molecules (Basel), 24 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service