Skip to Content
Merck
All Photos(2)

Key Documents

932698

Sigma-Aldrich

N4,N4,N4′,N4′-Tetra[(1,1′-biphenyl)-4-yl]-(1,1′-biphenyl)-4,4′-diamine

≥99% (HPLC)

Synonym(s):

N,N′-Tetra(4-biphenyl)benzidine, 4-Phenyl-n-(4-phenylphenyl)-n-[4-[4-(4-phenyl-n-(4-phenylphenyl)anilino)phenyl]phenyl]aniline, N,N,N′,N′-Tetrakis(4-biphenylyl)benzidine, N4,N4,N4′,N4′-Tetra(4-biphenylyl)-biphenyl-4,4′-diamine, BPBPA, [1,1′-Biphenyl]-4,4′-diamine, N4,N4,N4′,N4′-tetrakis([1,1′-biphenyl]-4-yl), [1,1′-Biphenyl]-4,4′-diamine, N4,N4,N4′,N4′-tetrakis([1,1′-biphenyl]-4-yl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H44N2
CAS Number:
Molecular Weight:
793.00
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21
grade:
sublimed grade
Pricing and availability is not currently available.

grade

sublimed grade

Quality Level

Assay

≥99% (HPLC)

SMILES string

N(c5ccc(cc5)c6ccc(cc6)N(c9ccc(cc9)c%10ccccc%10)c7ccc(cc7)c8ccccc8)(c3ccc(cc3)c4ccccc4)c1ccc(cc1)c2ccccc2

InChI

1S/C60H44N2/c1-5-13-45(14-6-1)49-21-33-55(34-22-49)61(56-35-23-50(24-36-56)46-15-7-2-8-16-46)59-41-29-53(30-42-59)54-31-43-60(44-32-54)62(57-37-25-51(26-38-57)47-17-9-3-10-18-47)58-39-27-52(28-40-58)48-19-11-4-12-20-48/h1-44H

InChI key

WXAIEIRYBSKHDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N4,N4,N4′,N4′-Tetra[(1,1′-biphenyl)-4-yl]-(1,1′-biphenyl)-4,4′-diamine, also known as BPBPA, is a Hole Transport, Electron Blocking Layer (HTL / EBL) material. BPBPA is popularly used in optoelectronic devices, such as high efficiency and long lifetime light-emitting diode devices (organic LEDs, OLEDs).
N4,N4,N4′,N4′-Tetra[(1,1′-biphenyl)-4-yl]-(1,1′-biphenyl)-4,4′-diamine is used as a hole transport material or as a host material in the emissive layer of organic light-emitting diodes (OLEDs) devices. Its semiconducting properties suits for R&D studies in organic field-effect transistors(OFETs) and thin-film transistors (TFTs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel molecular design employing a backbone freezing linker for improved efficiency, sharpened emission and long lifetime in thermally activated delayed fluorescence emitters
Yu, Ji Gwang et al.
Journal of Materials Chemistry, 7, 2919-2926 (2019)
High triplet energy exciplex host derived from a CN modified carbazole based n-type host for improved efficiency and lifetime in blue phosphorescent organic light-emitting diodes
Shin, Su Kyeong, et al.
Journal of Materials Chemistry, 6, 10308-10314 (2018)
Superb lifetime of blue organic light-emitting diodes through engineering interface carrier blocking layers and adjusting electron leakage and an unusual efficiency variation at low electric field
Kim, Taekyung et al.
Journal of Materials Chemistry, 6, 8472-8478 (2018)
Efficient Wide-Bandgap Mixed-Cation and Mixed-Halide Perovskite Solar Cells by Vacuum Deposition
Gil-Escrig, Lidon, et al.
ACS Energy Letters, 6, 827-836 (2021)
Do Sik Kim et al.
ACS applied materials & interfaces, 12(17), 19737-19745 (2020-04-18)
Four electron transport type hosts withstanding the positive polaron stress were synthesized using the benzo[4,5]thieno[3,2-d]pyrimidine core to develop high triplet energy hosts. Four benzo[4,5]thieno[3,2-d]pyrimidine-derived hosts, 4-(9H-carbazol-9-yl)-2-(3-(triphenylsilyl)phenyl)benzo[4,5]thieno[3,2-d]pyrimidine (CzBTPmSi), 2,4-di(9H-carbazol-9-yl)benzo[4,5]thieno[3,2-d]pyrimidine (2CzBTP), 2-(9H-carbazol-9-yl)-4-(3-(triphenylsilyl)phenyl)benzo[4,5]thieno[3,2-d]pyrimidine (mSiBTPCz), and 2,4-bis(3-(triphenylsilyl)phenyl)benzo[4,5]thieno[3,2-d]pyrimidine (2mSiBTP), were designed to have either the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service