Skip to Content
Merck
All Photos(2)

Documents

931063

Sigma-Aldrich

PEPPSI-IPr catalyst ChemBeads

Synonym(s):

[1,3-Bis(2,6-Diisopropylphenyl)imidazol-2-ylidene](3-chloropyridyl)palladium(II) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40Cl3N3Pd
CAS Number:
Molecular Weight:
679.46
UNSPSC Code:
12352103
NACRES:
NA.82

Assay

~5 wt.% loading of catalyst

Quality Level

form

solid

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

extent of labeling

~5 wt. % loading of catalyst

InChI

1S/C27H38N2.C5H4ClN.2ClH.Pd/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;6-5-2-1-3-7-4-5;;;/h9-16,18-21H,17H2,1-8H3;1-4H;2*1H;/q;;;;+2/p-2

InChI key

BLDKGTGQENJFON-UHFFFAOYSA-L

Application

PEPPSI-Ipr Catalyst is an air and temperature stable catalyst that is works well for Kumada-Tamao-Corriu (KTC), Negishi coupling, Suzuki coupling and Buchwald-Hartwig amination reactions. ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (669032)

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer’s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com

Patented, U.S. Pat. No. 7,250,510. Sold under an exclusive license from Total Synthesis Ltd.
PEPPSI is a trademark of Total Synthesis Ltd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PEPPSI palladium N-heterocyclic-carbene catalyst system enhances efficiency and functional group tolerance in catalysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service