Skip to Content
Merck
All Photos(1)

Documents

92855

Sigma-Aldrich

Glycerol trienanthate

≥94%

Synonym(s):

1,2,3-Trienanthoylglycerol, Glycerol triheptanoate, Triheptanoin, Trioenanthin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5COOCH2]2CHOCO(CH2)5CH3
CAS Number:
Molecular Weight:
428.60
Beilstein:
1807724
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥94% (TLC)
≥94%

form

liquid

refractive index

n20/D 1.444

density

0.966 g/mL at 20 °C (lit.)

SMILES string

CCCCCCC(OCC(COC(CCCCCC)=O)OC(CCCCCC)=O)=O

InChI

1S/C24H44O6/c1-4-7-10-13-16-22(25)28-19-21(30-24(27)18-15-12-9-6-3)20-29-23(26)17-14-11-8-5-2/h21H,4-20H2,1-3H3

InChI key

PJHKBYALYHRYSK-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

404.6 °F - open cup

Flash Point(C)

207 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan M Pascual et al.
JAMA neurology, 71(10), 1255-1265 (2014-08-12)
Disorders of brain metabolism are multiform in their mechanisms and manifestations, many of which remain insufficiently understood and are thus similarly treated. Glucose transporter type I deficiency (G1D) is commonly associated with seizures and with electrographic spike-waves. The G1D syndrome
Keri J Fogle et al.
Molecular genetics and metabolism, 126(4), 439-447 (2019-01-27)
Seizures are a feature not only of the many forms of epilepsy, but also of global metabolic diseases such as mitochondrial encephalomyopathy (ME) and glycolytic enzymopathy (GE). Modern anti-epileptic drugs (AEDs) are successful in many cases, but some patients are
Mohan Gomathi et al.
Journal of autism and developmental disorders, 50(8), 2740-2764 (2020-02-06)
Drug studies on Rett syndrome (RTT) have drastically increased over the past few decades. This review aims to provide master data on bench-to-bedside drug studies involving RTT. A comprehensive literature review was performed by searching in PUBMED, MEDLINE and Google
Sophie Calvert et al.
European journal of paediatric neurology : EJPN : official journal of the European Paediatric Neurology Society, 22(6), 1074-1080 (2018-08-22)
Despite antiepileptic medication and dietary treatment options available about 45% of children with epilepsy still suffer from uncontrolled seizures. Triheptanoin is an anaplerotic treatment designed to improve energy generation via the Krebs cycle. For the first time, we evaluated the
Mahmoud Reza Ashrafi et al.
Brain & development, 39(5), 369-385 (2017-01-25)
Childhood leukodystrophies are a growing category of neurological disorders in pediatric neurology practice. With the help of new advanced genetic studies such as whole exome sequencing (WES) and whole genome sequencing (WGS), the list of childhood heritable white matter disorders

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service