Skip to Content
Merck
All Photos(1)

Key Documents

924997

Sigma-Aldrich

E7820-C9-NH2 hydrochloride

Synonym(s):

N-(9-Aminononyl)-3-(N-(3-cyano-4-methyl-1H-indol-7-yl)sulfamoyl)benzamide hydrochloride, Crosslinker−E3 Ligase ligand conjugate, DCAF15 protein degrader building block

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€379.00

€379.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€379.00

About This Item

Empirical Formula (Hill Notation):
C26H33N5O3S · xHCl
Molecular Weight:
495.64 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.22

€379.00


Check Cart for Availability

Request a Bulk Order

ligand

E7820

Quality Level

reaction suitability

reactivity: carboxyl reactive
reagent type: ligand-linker conjugate

functional group

amine

Looking for similar products? Visit Product Comparison Guide

Application

Protein degrader building block E7820-C9-NH2 hydrochloride enables the synthesis of molecules for targeted protein degradation and PROTAC® (proteolysis-targeting chimeras) research. This conjugate contains a DCAF15-recruiting ligand E7820 (SML2950), an alkyl linker, and a pendant amine for reactivity with a carboxylic acid on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and degrader, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a terminal amine, parallel synthesis can be used to more quickly generate degrader libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.

Protein Degrader Building Blocks

Technology Spotlight: DCAF15 Degrader Building Blocks for Targeted Protein Degradation

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo target protein degradation induced by PROTACs based on E3 ligase DCAF15.
Liang Li et al.
Signal transduction and targeted therapy, 5(1), 129-129 (2020-07-28)
Tasuku Ishida et al.
SLAS discovery : advancing life sciences R & D, 26(4), 484-502 (2020-11-05)
Bifunctional degrader molecules, also called proteolysis-targeting chimeras (PROTACs), are a new modality of chemical tools and potential therapeutics to understand and treat human disease. A required PROTAC component is a ligand binding to an E3 ubiquitin ligase, which is then joined to another ligand binding to a protein to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service