Skip to Content
Merck
All Photos(1)

Key Documents

923850

Sigma-Aldrich

Alginate Aldehyde

20% aldehyde content, medium viscosity

Synonym(s):

Alginate aldehyde, Oxidized alginate

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€239.00

€239.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€239.00

About This Item

Linear Formula:
(C6H7NaO6)n(C6H3NaO6)m
UNSPSC Code:
12352201
NACRES:
NA.23

€239.00


Check Cart for Availability

Request a Bulk Order

Quality Level

description

Degree of functionalization: 15-25%

form

powder

color

white to off-white

suitability

conforms to structure for NMR

storage temp.

2-8°C

SMILES string

CO[C@H]1[C@@H](O)[C@H](O)[C@H](O/C(C=O)=C\C([O-])=O)O[C@@H]1C([O-])=O.OC2[C@H](OC(O)C=O)[C@@H](C([O-])=O)O[C@@H](OC)[C@H]2O

Looking for similar products? Visit Product Comparison Guide

Application

Alginate is an anionic polysaccharide that is widely used in pharmaceutical and biomedical applications due to its non-animal origin, low toxicity, biocompatibility, and biodegradability[1]. Alginate hydrogels are commonly used to fabricate tissue engineering scaffolds[2], bioinks for 3D bioprinting[3][4], and nanocarriers for drug and gene delivery.

Alginate is commonly crosslinked into a hydrogel via ionic-crosslinking with divalent cations (e.g., Ca2+). To prevent matrix degradation, alginate can be functionalized with reactive groups that can be chemically crosslinked, such as aldehydes.[5] Aldehyde- functionalized alginate can be used to prepare hydrogels by reaction with amine groups, such as gelatin through Schiff-base reaction to form a chemical hydrogel. This material can be used in a variety of biomedical applications such as the delivery of drugs, cells, or biomolecules in different tissues, wound healing, and muscle and bone tissue engineering. [6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Degree of Substitution of Formyl Groups in Polyaldehyde Dextran by the Hydroxylamine Hydrochloride Method
Huiru Zhao, Ned D. Heindel
Pharmaceutical Research, 8, 400?402-400?402 (1991)
The effect of oxidation on the degradation of photocrosslinkable alginate hydrogels
Jeon O, et al.
Biomaterials, 33(13), 3503-3514 (2012)
Biji Balakrishnan et al.
Biomaterials, 26(18), 3941-3951 (2005-01-01)
The injectable polymer scaffolds which are biocompatible and biodegradable are important biomaterials for tissue engineering and drug delivery. Hydrogels derived from natural proteins and polysaccharides are ideal scaffolds for tissue engineering since they resemble the extracellular matrices of the tissue
Controlling alginate oxidation conditions for making alginate-gelatin hydrogels
Emami Z, et al.
Carbohydrate Polymers, 198, 509-517 (2018)
B Balakrishnan et al.
Carbohydrate research, 340(7), 1425-1429 (2005-04-28)
Periodate oxidation of sodium alginate in aqueous solution as well as a dispersion in 1:1 ethanol-water was examined. The oxidation proceeded smoothly in both media, and the kinetics of oxidation was surprisingly similar. Polymer cleavage was observed in both media

Questions

  1. Is the alginate aldehyde terminated, as described in the linear formula, with only 20% of the chains functionalized? Or is the backbone of the chain functionalized with aldehyde with 20% of the monomers containing aldehyde groups?

    1 answer
    1. Only the aldehyde content is determined by the titration with hydroxylamine. The location of the aldehyde groups is not determined and the image is representative.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service