Skip to Content
Merck
All Photos(1)

Key Documents

923427

Sigma-Aldrich

2-(4-((2-(tert-Butoxycarbonyl)-2-azaspiro[3.3]heptan-6-yl)oxy)piperidin-1-yl)acetic acid

Synonym(s):

Rigid linker for PROTAC® development

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€360.00

€360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C18H30N2O5
Molecular Weight:
354.44
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

€360.00


Check Cart for Availability

Request a Bulk Order

form

powder or crystals

Quality Level

reaction suitability

reagent type: linker

functional group

Boc
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(CN1CCC(OC2CC3(CN(C(OC(C)(C)C)=O)C3)C2)CC1)=O

InChI

1S/C18H30N2O5/c1-17(2,3)25-16(23)20-11-18(12-20)8-14(9-18)24-13-4-6-19(7-5-13)10-15(21)22/h13-14H,4-12H2,1-3H3,(H,21,22)

InChI key

FBAXECBIBBBIPB-UHFFFAOYSA-N

Application

2-(4-((2-(tert-Butoxycarbonyl)-2-azaspiro[3.3]heptan-6-yl)oxy)piperidin-1-yl)acetic acid is useful as a rigid linker in PROTAC development for targeted protein degradation as well as chemical conjugates. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties. Developed in partnership with ComInnex.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service