Skip to Content
Merck
All Photos(1)

Key Documents

92251

Sigma-Aldrich

Trimethylamine

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein:
956566
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

2.09 (vs air)

vapor pressure

13.3 psi ( 21 °C)

Assay

≥99.0%

form

gas

autoignition temp.

374 °F

expl. lim.

11.6 %

impurities

≤0.05% ammonia
≤0.05% methylamine
≤0.2% dimethylamine

bp

3-4 °C (lit.)

mp

−117 °C (lit.)

density

0.63 g/mL at 20 °C (lit.)

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin with 250 mL (net ~155 g)

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E L Barrett et al.
Annual review of microbiology, 39, 131-149 (1985-01-01)
Trimethylamine oxide, which is found in relatively high concentrations in the tissues of marine animals, serves as an electron acceptor in the anaerobic metabolism of a number of bacteria associated primarily with three environments: the marine environment (e.g. Alteromonas and
M Al-Waiz et al.
Clinical pharmacology and therapeutics, 42(5), 588-594 (1987-11-01)
Trimethylamine (TMA) and its N-oxide (TMAO) are normal components of human urine. They are present in the diet and also derived from the enterobacterial metabolism of precursors such as choline. Dietary TMA is almost entirely metabolized to and excreted as
S L Hillier
American journal of obstetrics and gynecology, 169(2 Pt 2), 455-459 (1993-08-01)
Bacterial vaginosis is characterized microbiologically by replacement of the Lactobacillus-predominant vaginal flora by Gardnerella vaginalis, Bacteroides species, Mobiluncus species, and genital mycoplasmas. A standardized, laboratory-based diagnostic test for bacterial vaginosis is desirable in those instances in which a microscope is
Hyeryeong Jeon et al.
Macromolecular rapid communications, 33(11), 972-976 (2012-04-12)
Polydiacetylenes (PDAs), a family of conjugated polymers, are very intriguing materials in several aspects. Especially, the stimulus-induced apparent blue-to-red transition of the PDAs has led to the development of a variety of PDA-based chemosensors. In the current work, we synthesized
Smaranda Craciun et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(52), 21307-21312 (2012-11-16)
Choline and trimethylamine (TMA) are small molecules that play central roles in biological processes throughout all kingdoms of life. These ubiquitous metabolites are linked through a single biochemical transformation, the conversion of choline to TMA by anaerobic microorganisms. This metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service