Skip to Content
Merck
All Photos(5)

Key Documents

922196

Sigma-Aldrich

Mini Luer to pipette adapter

Fluidic 391, PP

Synonym(s):

Microfluidics accessory

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€58.90

€58.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€58.90

About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

€58.90


Check Cart for Availability

Request a Bulk Order

Application

The Mini Luer connector can be used to connect microfluidic platforms with tubing or directly with instrumentation. The Mini Luer to pipette adapters allow a flush sealing of a pipette tip to a chip equipped with a Mini Luer interfaces. This enables the realization of higher applied fluidic pressures, as well as a reduced contamination risk. The Mini Luer connector is made of hard polypropylene (PP). Avoid force on the PP interface and chip to prevent chip damage.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances of controlled drug delivery using microfluidic platforms.
Li X, et al.
Advanced Drug Delivery Reviews, 128, 3-28 (2018)
Microfluidic-assisted fabrication of carriers for controlled drug delivery.
Santos H A, et al.
Lab on a chip, 17, 1856-1883 (2017)
Recent advances of controlled drug delivery usingmicrofluidic platforms.
Li X, et al.
Advanced Drug Delivery Reviews, 128, 3-28 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service