Skip to Content
Merck
All Photos(1)

Key Documents

921599

Sigma-Aldrich

ZnO nanoparticle ink solution

2.5 wt. %, viscosity 2.8 cP , work function 4.3 eV

Synonym(s):

Ink for OLED, Ink for charge selective layers in printed electronics, OPV, PV, Photodetectors, QLED, Zinc oxide ink, Zinc oxide nanoparticle dispersion

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€107.00

€107.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€107.00

About This Item

Empirical Formula (Hill Notation):
ZnO
CAS Number:
Molecular Weight:
81.39
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

€107.00


Check Cart for Availability

Request a Bulk Order

form

suspension

Quality Level

concentration

2.5 wt. %

work function

4.3 eV

viscosity

2.8 cP

SMILES string

[Zn]=O

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This zinc oxide nanoparticle ink is a nanoparticle-based printing ink allowing processing temperatures of 80-120 °C. The zinc oxide nanoparticle ink is an electron selective interface layer ink containing zinc oxide (ZnO) nanoparticles in butanols. ZnO nanoparticle ink functions well as electron extraction layer (EEL) materials in solar cells.

Application

This ZnO nanoparticle ink is for slot-die, spin-coating, doctor blading or inkjet printing for the use as electron transport layer in printed electronics. The ZnO nanoparticle ink is universally applicable in normal and inverted architecture.
Annealing temperature 80-120 °C.

Preparation Note

  • Storage: In dark at room temperature
  • Prior to application: Shake, ultrasonicate and (optionally) filter through 0.45 μm PTFE filter
  • Post-treatment: Annealing of deposited ZnO films at 80-120 °C
  • Thickness optimization may be necessary

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

83.3 °F

Flash Point(C)

28.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Find advantages of inorganic interface layer inks for organic electronic & other applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service