Skip to Content
Merck
All Photos(5)

Key Documents

920940

Sigma-Aldrich

Monolayer hexagonal Boron Nitride (hBN) on Si/SiO2 wafer

diam. 150 mm (6 in.)

Synonym(s):

Boronitrene, Hexagonal boron nitride monolayer, Single-layer hexagonal boron nitride, White graphene

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€942.00

€942.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€942.00

About This Item

Empirical Formula (Hill Notation):
BN
CAS Number:
Molecular Weight:
24.82
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.23

€942.00


Check Cart for Availability

Request a Bulk Order

description

Bandgap: 5.97 eV
Metal Impurities: 1.00e10 – 5.00e10 (at/cm2)
Orientation: <1-0-0>
Raman Peak: 1370 /cm-1
Substrate Type/Doping: P/B
hBN Coverage: 100% with sporadic adlayers
1-25 Ω-cm

Quality Level

diam.

150 mm (6 in.)

size

300 nm , Oxide

thickness

675 ± 25 μm , Wafer

grain size

>4 μm

SMILES string

B#N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monolayer hexagonal boron nitride (h-BN), also known as “white graphene”, is a wide-bandgap 2D crystal (~6 eV that can be tuned to ~2 eV) [1][2] with exceptional strength [3], large oxidation resistance at high temperatures [3], and optical functionalities [2][3]. Among its potential applications are:
  • Two-dimensional electronics [1][2][3]
  • Nanophotonic and other optoelectronic devices †††
  • Quantum communication and information science [2]
  • Aerospace industry [4]
  • MEMS and NEMS [5]
  • Micro-/nano- actuators [3]
  • Insulating/transparent coatings [1]

Storage and Stability

To ensure the maximum shelf life of your hBN sample, it is best stored under vacuum or in inert atmosphere (Argon or Nitrogen) conditions once the vacuum sealed package has been opened.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hexagonal boron nitride nanomechanical resonators with spatially visualized motion
Zheng X, et al.
Microsystems & Nanoengineering, 3 (2017)
From 2-D to 0-D Boron Nitride Materials, The Next Challenge
Stagi L, et al.
Materials, 12(23) (2019)
Monolayer to Bulk Properties of Hexagonal Boron Nitride
Wickramaratne D, et al.
Physical Chemistry, 122, 25524-25529 (2018)
Chemical and Bandgap Engineering in Monolayer Hexagonal Boron Nitride
Ba K, et al.
Scientific reports, 7 (2017)
Piezoelectricity in Monolayer Hexagonal Boron Nitride
Ares P, et al.
Advanced Materials, 32 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service