Skip to Content
Merck
All Photos(1)

Key Documents

920347

Sigma-Aldrich

Lithium hexafluoroarsenate(V)

anhydrous, 99.9% trace metals basis

Synonym(s):

LiAsF6, Lithium fluoarsenate, Hexafluoroarsenate(V) lithium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAsF6
CAS Number:
Molecular Weight:
195.85
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
anhydrous
battery grade
form:
powder
Pricing and availability is not currently available.

grade

anhydrous
battery grade

Quality Level

Assay

99.9% trace metals basis

form

powder

impurities

≤100 ppm H2O

mp

240 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Li+].F[As-](F)(F)(F)(F)F

InChI

1S/AsF6.Li/c2-1(3,4,5,6)7;/q-1;+1

InChI key

GTZQZEYBOGZTEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hexafluoroarsenate(V) anhydrous, 99.9% trace metals basis is a highly pure (99.9%) with ≤100 ppm H2O, is designed to support research in battery. It comes as a white color powder/crystal with trace metal ≤ 1500.0 ppm.

Application

Lithium hexafluoroarsenate(V) (LiAsF6) is primarily used as an electrolyte salt in lithium-ion batteries, where it enhances performance, stability, and energy density. It can be used in the development of electrolytes for supercapacitors, where it helps improve the charge-discharge cycles and energy storage capacity. Additionally, it is used in electrochemical devices, advanced material synthesis, catalysis, research and development, environmental applications, and analytical chemistry.[1][2][3]
The surface chemistry of Li electrodes in ethereal LiAsF6 was tested with new salts for potential battery systems.

Features and Benefits

The anhydrous form of lithium hexafluoroarsenate(V), with a 99.9% trace metals basis, is well-suited for use in the rechargeable energy sector. Its meticulous control of parameters, including trace metal impurities and water content, ensures dependable results in high-end experiments.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Interplay between Salt Association and the Dielectric Properties of Low Permittivity Electrolytes: The Case of LiPF6 and LiAsF6 in Dimethyl Carbonate.
Self J, et al.
The Journal of Physical Chemistry C, 122(4), 1990-1994 (2018)
Structure, Electrical and Optical Properties of (PVA/LiAsF6) Polymer Composite Electrolyte Films
Varishetty M M , et al
Polym. Eng. Sci., 50, 878-884 (2010)
Guided Lithium Metal Deposition and Improved Lithium Coulombic Efficiency through Synergistic Effects of LiAsF6 and Cyclic Carbonate Additives.
Ren X, et al.
ACS Energy Letters, 3(1), 14-19 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service