917826
Indium tin oxide coated PEN
80% transparency, L × W × thickness 29.7 cm × 21 cm × 125 μm, sheet resistance <15 Ω/sq
Synonym(s):
ITO coated polyethylene naphthalate, ITO-PEN (80% transparency), PECF-IP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
substrate (Polyethylene naphthalate (PEN) film 125 micron thick (one side coated))
Quality Level
sheet resistance
<15 Ω/sq
L × W × thickness
29.7 cm × 21 cm × 125 μm
transmittance
550 nm, 80%
Application
This product is a one-side coated, highly transparent, flexible, ITO coated PEN polymer film. It is a ready-to-use flexible, transparent conducting substrate with low sheet resistance.
This product can be used for rapid fabrication of flexible electronics such as touch screen, Perovskite solar cells, sensors, and dye-sensitized solar cells.
This product can be used for rapid fabrication of flexible electronics such as touch screen, Perovskite solar cells, sensors, and dye-sensitized solar cells.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 2 Inhalation
Target Organs
Lungs
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Langmuir : the ACS journal of surfaces and colloids, 30(31), 9397-9406 (2014-07-17)
In this paper, we report a simple and versatile process of electrografting the aryl multilayers onto indium tin oxide (ITO)-coated flexible poly(ethylene naphthalate) (PEN) substrates using a diazonium salt (4-pyrrolylphenyldiazonium) solution, which was generated in situ from a reaction between
Dependence of ITO-Coated Flexible Substrates in the Performance and Bending Durability of Perovskite Solar Cells.
Advanced Engineering Materials, 21(8), 1900288-1900288 (2019)
Photovoltaic Performance of Plastic Dye-Sensitized Electrodes Prepared by Low-Temperature Binder-Free Coating of Mesoscopic Titania.
Journal of the Electrochemical Society, 154(5) (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service