Skip to Content
Merck
All Photos(1)

Key Documents

91733

Sigma-Aldrich

Trimethylsilyl trifluoroacetate

≥97.0% (T)

Synonym(s):

Trifluoroacetic acid trimethylsilyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CO2Si(CH3)3
CAS Number:
Molecular Weight:
186.20
Beilstein:
1773314
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

≥97.0% (T)

form

liquid

refractive index

n20/D 1.336 (lit.)
n20/D 1.336

bp

88-90 °C (lit.)

density

1.078 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

C[Si](C)(C)OC(=O)C(F)(F)F

InChI

1S/C5H9F3O2Si/c1-11(2,3)10-4(9)5(6,7)8/h1-3H3

InChI key

VIYXXANHGYSBLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For the ring opening of cyclopropyl ketones[1]; XeF2-mediated trifluoromethylation of aromatic compounds[2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie A Balko et al.
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians, 49(3), 680-688 (2018-09-14)
Prolonged anesthetic recovery time is a common complication of chelonian inhalant anesthesia and may be exacerbated by right-to-left intracardiac shunting of blood. Epinephrine may decrease intracardiac shunting, which may shorten anesthetic recovery time. The study objective was to assess inhalant
Y. Tanabe et al.
The Journal of Organic Chemistry, 53, 4582-4582 (1988)
M. Demuth et al.
Helvetica Chimica Acta, 64, 2759-2759 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service