Skip to Content
Merck
All Photos(1)

Key Documents

917273

Sigma-Aldrich

Tapered dispensing tips

20G, 0.50″, 50/box

Synonym(s):

3d bioprinting consumables, Bioprinting nozzle

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€68.30

€68.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€68.30

About This Item

UNSPSC Code:
42182011
NACRES:
NB.51

€68.30


Check Cart for Availability

Request a Bulk Order

Quality Level

description

Tapered dispensing tips, polypropylene, 20 gauge, pink, ID 0.023″, 50/box

Application

The tapered design provides smooth flow of viscous fluids and resists clogging when dispensing particle-filled materials.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service