Skip to Content
Merck
All Photos(1)

Documents

916234

Sigma-Aldrich

5,5-Dimethyl-2-phenoxy-1,3,2-dioxaphosphorinane

≥95%

Synonym(s):

2,2-Dimethyl trimethylene phenyl phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15O3P
CAS Number:
Molecular Weight:
226.21
MDL number:
UNSPSC Code:
12352200

Assay

≥95%

form

liquid

storage temp.

2-8°C

Application

5,5-Dimethyl-2-phenoxy-1,3,2-dioxaphosphorinane is a phosphite ligand for Pd catalysis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service