Skip to Content
Merck
All Photos(2)

Key Documents

915556

Sigma-Aldrich

HNMB

≥98%

Synonym(s):

4-[(E)-[(2-hydroxy-1-naphthalenyl)methylene]amino]-3-methyl benzoic acid, Aggregation-induced emmision (AIE) materials, Mechanochromic luminescent (MCL) materials

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€160.00

€160.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€160.00

About This Item

Empirical Formula (Hill Notation):
C19H15NO3
CAS Number:
Molecular Weight:
305.33
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€160.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98%

form

solid

color

white

mp

292.9 °C

UV absorption

λ: 317 nm Amax
λ: 443 nm Amax

SMILES string

N(=C\c2c3c(ccc2O)cccc3)/c1c(cc(cc1)C(=O)O)C

InChI

1S/C19H15NO3/c1-12-10-14(19(22)23)6-8-17(12)20-11-16-15-5-3-2-4-13(15)7-9-18(16)21/h2-11,21H,1H3,(H,22,23)/b20-11+

InChI key

HCGGUBXKKWXERU-RGVLZGJSSA-N

Application

HNMB is a mechanochromic luminescent material with aggregation-induced emission (AIE). It has applications for pressure sensing and mapping.
In biology, pressure could direct the folding-unfolding of protein molecules, guide blood stream and even control the cell migration. The complexity with these cases makes the monitor and measurement of pressure from conventional force sensor based on classical mechanics impractical. Based on mechanical sensitive materials with ingenious design at the molecular level, a new way to detect pressure is developed. And mechanochromic luminescent (MCL) materials have formed a vital part of such materials. They are intentionally endowed with the ability to change luminescence property (e.g., emission wavelength, emission intensity and lifetime) under mechanical force, which can be readily detected by naked eyes or instruments. As molecule-based sensors, they are compatible to various flexible substrates and arbitrarily curvilinear surfaces, facilitating many applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eethamukkala Ubba et al.
Chemistry, an Asian journal, 13(21), 3106-3121 (2018-08-10)
Mechanoluminescence (ML) involves the emission of light from various organic and inorganic materials upon mechanical stimulus. In this review, we contemplate the advances of metal-free, purely organic ML compounds with aggregation-induced emission (AIE), which give strong emission upon mechanical force.
Mengyao Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 220, 117125-117125 (2019-05-29)
In this study, we report a new compound, (E)-4-(((2-hydroxynaphthalen-1-yl)methylene)amino)-3-methylbenzoic acid (HNMB), which shows aggregation-induced emission property as well as intramolecular charge transfer (ICT) nature. In addition, it exhibits unique mechanochromic luminescence (MCL). The HNMB solid powder emits strong emission but
Xin Qian et al.
Advanced materials (Deerfield Beach, Fla.), 30(25), e1800291-e1800291 (2018-05-04)
Mechanically driven light generation is an exciting and under-exploited phenomenon with a variety of possible practical applications. However, the current driving mode of mechanoluminescence (ML) devices needs strong stimuli. Here, a flexible sensitive ML device via nanodopant elasticity modulus modification
Trap-controlled mechanoluminescent materials.
Zhang J C, et al.
Progress in Materials Science, 103, 678-742 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service