Skip to Content
Merck
All Photos(1)

Key Documents

915254

Sigma-Aldrich

ZrO2 nanoparticles

5 nm (core), acrylate functionalized, 50 wt. % in propylene glycol methyl ether acetate (PGMEA), medium surface coverage capping material

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€404.00

€404.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€404.00

About This Item

Linear Formula:
ZrO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

€404.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid

concentration

50 wt. % in propylene glycol methyl ether acetate (PGMEA)

particle size

5 nm (core)

functional group

acrylate

SMILES string

[Zr](=O)=O

InChI

1S/2O.Zr

InChI key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High refractive index nanocrystal dispersions dramatically improve optical and mechanical properties to enable next-generation products. Incorporating ZrO2 significantly increases the refractive index of many commonly used polymers, while maintaining high transparency. This combination of key properties allows researchers to optimize optical efficiency and improve light extraction.

Features and Benefits

  • Zirconia with 5nm core plus surface capping that contains acrylate functionality designed to crosslink with acrylate and vinyl monomers.
  • Balanced surface coverage that provides both improved formulation stability and dispersibility without sacrificing refractive index.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F

Flash Point(C)

46 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Target applications for the ZrO2 include AR/VR, optical devices such as diffractive optical elements, wafer-level optics, sensors, and micro-lens arrays, and various display technologies including organic light-emitting diodes, quantum dots, and mini and micro light-emitting diodes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service