Skip to Content
Merck
All Photos(2)

Key Documents

914878

Sigma-Aldrich

Silica

mesoporous, 200 nm particle size, pore size ~2 nm

Synonym(s):

Porous Silica, Silicon dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€150.00

€150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€150.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

€150.00


Check Cart for Availability

Request a Bulk Order

form

mesoporous

Quality Level

particle size

200 nm

pore size

~2 nm

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mesoporous silicas are inorganic materials having porous structures and find applications in chromatography; catalysis; drug delivery and imaging.[1][2][3]

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Wang et al.
Advanced materials (Deerfield Beach, Fla.), 27(3), 576-585 (2014-09-23)
Advances in nanotechnology and nanomedicine offer great opportunities for the development of nanoscaled theranostic platforms. Among various multifunctional nanocarriers, magnetic mesoporous silica nanocomposites (M-MSNs) attract prominent research interest for their outstanding properties and potential biomedical applications. This Research News article
Si-Han Wu et al.
Chemical Society reviews, 42(9), 3862-3875 (2013-02-14)
Good control of the morphology, particle size, uniformity and dispersity of mesoporous silica nanoparticles (MSNs) is of increasing importance to their use in catalyst, adsorption, polymer filler, optical devices, bio-imaging, drug delivery, and biomedical applications. This review discusses different synthesis
Isabel Sierra et al.
Journal of chromatography. A, 1363, 27-40 (2014-07-13)
Recent advances in the development of new materials are having a major impact on analytical chemistry. For example, the unique properties of ordered mesoporous silicas (OMSs) have been shown to enhance the analytical performance of many existing techniques or allow

Questions

  1. can you please told me about the charge (means negative or Positive charge on the surface of silica nanoparticles) on this silica nanopartilces and does it have secondary amine group in the structure?

    1 answer
    1. The charge information is not determined for this product. This product does not contain a secondary amine group in the structure.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service