Skip to Content
Merck
All Photos(2)

Key Documents

914339

Sigma-Aldrich

Fmoc-L-Lys(Nvoc)-OH

≥98%

Synonym(s):

(S)-2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)-6-((4,5-dimethoxy-2-nitrobenzyloxy)carbonylamino)hexanoic acid, N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-(((4,5-dimethoxy-2-nitrobenzyl)oxy)carbonyl)-L-lysine, Lysine with photoremovable NVOC, Photocaged amino acid, Photocleavable lysine derivative

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€279.00

€279.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€279.00

About This Item

Empirical Formula (Hill Notation):
C31H33N3O10
CAS Number:
Molecular Weight:
607.61
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€279.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

mp

108 °C (decomp.)

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1c(cc(c(c1)OC)OC)COC(=O)NCCCC[C@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O

InChI

1S/C31H33N3O10/c1-41-27-15-19(26(34(39)40)16-28(27)42-2)17-43-30(37)32-14-8-7-13-25(29(35)36)33-31(38)44-18-24-22-11-5-3-9-20(22)21-10-4-6-12-23(21)24/h3-6,9-12,15-16,24-25H,7-8,13-14,17-18H2,1-2H3,(H,32,37)(H,33,38)(H,35,36)/t25-/m0/s1

InChI key

UOEGMVQGRMCJDM-VWLOTQADSA-N

Application

Fmoc-L-Lys(Nvoc)-OH is a lysine derivative compatible with solid-phase peptide synthesis (SPPS). The photolabile, N-terminal protecting group nitroveratryloxycarbonyl (NVOC) can be cleaved by photolysis at 350 nm. Photocaged amino acids such as these are useful in the synthesis of photocleavable chemical tools for spatial and temporal control over released molecules in biological applications.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service