Skip to Content
Merck
All Photos(1)

Key Documents

91377

Sigma-Aldrich

Boron trifluoride acetonitrile complex solution

15.2-16.8% BF3 basis

Synonym(s):

(Acetonitrile)-trifluoroborane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€117.00

€117.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€117.00

About This Item

Empirical Formula (Hill Notation):
C2H3BF3N
CAS Number:
Molecular Weight:
108.86
Beilstein:
3910569
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€117.00


Estimated to ship on30 May 2025


Request a Bulk Order

form

liquid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

15.2-16.8% boron trifluoride

density

0.87-0.88 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CC#N.FB(F)F

InChI

1S/C2H3N.BF3/c1-2-3;2-1(3)4/h1H3;

InChI key

MEMUCXUKCBNISQ-UHFFFAOYSA-N

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service