Skip to Content
Merck
All Photos(3)

Key Documents

912956

Sigma-Aldrich

(2R,2′R,3R,3′R)-MeO-BIBOP

Synonym(s):

(2R,2′R,3R,3′R)-3,3′-Di-tert-butyl-4,4′-dimethoxy-2,2′,3,3′-tetrahydro-2,2′-bibenzo[d][1,3]oxaphosphole

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€239.00

€239.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€239.00

About This Item

Empirical Formula (Hill Notation):
C24H32O4P2
CAS Number:
Molecular Weight:
446.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

€239.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

optical purity

ee: ≥99% (HPLC)

reaction suitability

reagent type: ligand

functional group

phosphine

InChI key

KVPCVNGTJXEQKH-IKTNGCAPSA-N

Related Categories

Application

(2R,2′R,3R,3′R)-MeO-BIBOP is a P-chiral biphosphorus ligand used in a variety of asymmetric transition metal-catalyzed transformations including hydrogenations, propargylations, reductions, and hydroformylations.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

Sold in collaboration with Zejun Pharmaceuticals

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel R Fandrick et al.
Journal of the American Chemical Society, 132(22), 7600-7601 (2010-05-21)
The highly enantio- and regioselective copper catalyzed asymmetric propargylation of aldehydes with a propargyl borolane reagent is reported. The methodology demonstrated broad functional group tolerance and provided high enantioselectivities for aliphatic, vinyl, and aryl aldehydes. The utility of the TMS
Renchang Tan et al.
Organic letters, 18(14), 3346-3349 (2016-06-23)
Air-stable and tunable chiral bisdihydrobenzooxaphosphole ligands (BIBOPs) were employed in rhodium-catalyzed asymmetric hydroformylation of various terminal olefins with excellent conversions (>99%), moderate-to-excellent enantioselectivities (up to 95:5 er), and branched to linear ratios (b:l) of up to 400.
Chengxi Li et al.
Angewandte Chemie (International ed. in English), 58(38), 13573-13583 (2019-07-26)
We herein report the development of a conformationally defined, electron-rich, C2 -symmetric, P-chiral bisphosphorus ligand, ArcPhos, by taking advantage of stereoelectronic effects in ligand design. With the Rh-ArcPhos catalyst, excellent enantioselectivities and unprecedentedly high turnovers (TON up to 10 000) were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service