Skip to Content
Merck
All Photos(2)

Key Documents

912751

Sigma-Aldrich

Y5

≥99%

Synonym(s):

(2,2’-((2Z,2’Z)-((12,13-bis(2-ethylhexyl)-3,9-diundecyl-12,13-dihydro[1,2,5]thiadiazolo[3,4e]thieno[2’’,3’’:4’,5’] thieno[2’,3’:4,5]pyrrolo[3,2-g] thieno[2’,3’:4,5]thieno[3,2-b]indole-2,10-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro1H-indene-2,1-diylidene))dimalononitrile)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€478.00
250 MG
€956.00

€478.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€478.00
250 MG
€956.00

About This Item

Empirical Formula (Hill Notation):
C82H90N8O2S5
CAS Number:
Molecular Weight:
1379.97
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

€478.00


Check Cart for Availability

Request a Bulk Order

description

Band gap: Eg=1.38 eV

Quality Level

Assay

≥99%

form

solid

color

dark

Orbital energy

HOMO -5.55 eV 
LUMO -3.87 eV 

SMILES string

[s]1nc2c(n1)c3c4[s]c5c([s]c(c5CCCCCCCCCCC)\C=C%11/C(=O)c%12c(cccc%12)C/%11=C(C#N)C#N)c4[n](c3c6[n](c7c([s]c8c7[s]c(c8CCCCCCCCCCC)\C=C9\C(=O)c%10c(cccc%10)C\9=C(C#N)C#N)c62)CC(CCCC)CC)CC(CCCC)CC

InChI key

AQGSDAUCGRTLKJ-AXQTZOLBSA-N

General description

Non-fullerene acceptors (NFAs) are currently a major focus of research in the development of bulk-heterojunction organic solar cells (OSCs). In contrast to the widely used fullerene acceptors (FAs), the optical properties and electronic energy levels of NFAs can be designed and readily tuned. NFA-based OSCs can also achieve greater thermal stability and photochemical stability, as well as longer device lifetimes, than their FA-based counterparts. Recent developments have led to a rapid increase in power conversion efficiencies for NFA OSCs, with values now exceeding 15% in a single junction cell, and >17% for a tandem cell, demonstrating the viability of using NFAs to replace FAs in next-generation high-performance OSCs.

Application

Y5 is a high-performance non-fullerene acceptor (NFA) that uses an electron-deficient molecular core to obtain a low bandgap NFA and high absorption coefficient. OPV devices made from Y5 can reach high performances in single-junction devices. Y5 is versatile and can be employed in both conventional and inverted OPV device architectures and has reported PCE from 10.5 to 14.1% when paired with five different donor polymers.[1]
Y5 is a highly efficient low band-gap non-fullerene acceptor. OPV devices made from Y5 can reach high performances in single-junction devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Yuan et al.
Advanced materials (Deerfield Beach, Fla.), 31(17), e1807577-e1807577 (2019-03-19)
Narrow bandgap n-type organic semiconductors (n-OS) have attracted great attention in recent years as acceptors in organic solar cells (OSCs), due to their easily tuned absorption and electronic energy levels in comparison with fullerene acceptors. Herein, a new n-OS acceptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service