Skip to Content
Merck
All Photos(1)

Key Documents

91040

Sigma-Aldrich

Glyceryl trioctanoate

technical, ≥90% (GC)

Synonym(s):

1,2,3-Tricapryloylglycerol, 1,2,3-Trioctanoylglycerol, Glycerol tricaprylate, Glycerol trioctanoate, Glyceryl tricaprylate, Tricaprylin, Trioctanoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)6COOCH2]2CHOCO(CH2)6CH3
CAS Number:
Molecular Weight:
470.68
Beilstein:
1717202
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

form

liquid

bp

233 °C/1 mmHg (lit.)

density

0.956 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CCCCCCCC(=O)OCC(COC(=O)CCCCCCC)OC(=O)CCCCCCC

InChI

1S/C27H50O6/c1-4-7-10-13-16-19-25(28)31-22-24(33-27(30)21-18-15-12-9-6-3)23-32-26(29)20-17-14-11-8-5-2/h24H,4-23H2,1-3H3

InChI key

VLPFTAMPNXLGLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong Ho Jeon et al.
Applied microbiology and biotechnology, 81(5), 865-874 (2008-09-06)
To search for new cold-active lipases, a metagenomic library was constructed using cold-sea sediment samples at Edison Seamount and was screened for lipolytic activities by plating on a tricaprylin medium. Subsequently, a fosmid clone was selected, and the whole sequence
Tehila Mishraki et al.
Colloids and surfaces. B, Biointerfaces, 75(1), 47-56 (2009-09-15)
A model protein (lysozyme) was incorporated into monoolein-based reverse hexagonal (H(II)) mesophase and its structure effects were characterized by small angle X-ray scattering, ATR-FTIR spectroscopy, and rheological measurements. Modifications in molecular organization of the H(II) mesophases as well as the
Maciej Pilarek et al.
Journal of biotechnology, 127(4), 736-744 (2006-09-30)
A new model of enzymatic 1,3-specific alcoholysis of triacylglycerols has been developed. The irreversibility of the acyl bounds cleavage in glycerides, a reversible monoglycerides isomerization and an irreversible enzyme deactivation have been assumed. The Ping Pong Bi Bi mechanism with
Ahmed Aloulou et al.
Biochimica et biophysica acta, 1771(12), 1446-1456 (2007-11-21)
The effects of various detergents and pH on the interfacial binding and activity of two fungal lipases from Yarrowia lipolytica (YLLIP2) and Thermomyces lanuginosus (TLL) were investigated using trioctanoin emulsions as well as monomolecular films spread at the air-water interface.
Pedro Reis et al.
Langmuir : the ACS journal of surfaces and colloids, 24(14), 7400-7407 (2008-06-13)
Tensiometry (the pendant drop technique), interfacial shear rheology, and ellipsometry have been used to study the effect of polar lipids that are generated during fat digestion on the behavior of lipases at the oil-water interface. Both Sn-1,3 regiospecific and nonregiospecific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service