Skip to Content
Merck
All Photos(2)

Key Documents

909610

Sigma-Aldrich

BMSO crosslinker

≥95%

Synonym(s):

3,3′-Sulfinylbis(N-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)propanamide), Mass spectrometry-cleavable crosslinker for studying protein-protein interations

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€298.00

€298.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€298.00

About This Item

Empirical Formula (Hill Notation):
C18H22N4O7S
CAS Number:
Molecular Weight:
438.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

€298.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

[S](=O)(CCC(=O)NCCN2C(=O)C=CC2=O)CCC(=O)NCCN1C(=O)C=CC1=O

InChI key

PUNDHDZIOGBGHG-UHFFFAOYSA-N

Application

BMSO (bismaleimide sulfoxide) crosslinker is a homobifunctional, cysteine residue-targeting, sulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS). BMSO possesses two maleimide reactive groups for targeting Cys, a 24.2 Å spacer arm, and two symmetrical C-S cleavable bonds adjacent to the central sulfoxide. While similar reagents react with lysine (Lys) and acidic residues, targeting Cys provides further opportunity to characterize protein interaction surfaces. Additionally, the post-cleavage spacer yields tagged peptides for unambiguous identification by collision-induced dissociation in tandem MS.

BMSO crosslinker provides complementary data to amine-reactive and acidic residue-targeting reagents and will find wide utility in the elucidation of PPIs, study of proteins that function as complexes, quantification of structural dynamics, and the quest for targeting "undruggable" protein targets.

Legal Information

Subject to US provisional patent application #62/845,240 of the Regents of the University of California

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig B Gutierrez et al.
Analytical chemistry, 90(12), 7600-7607 (2018-05-25)
Cross-linking mass spectrometry (XL-MS) has become an emerging technology for defining protein-protein interactions (PPIs) and elucidating architectures of large protein complexes. Up to now, the most widely used cross-linking reagents target lysines. Although such reagents have been successfully applied to
Craig Gutierrez et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(8), 4088-4098 (2020-02-09)
The COP9 signalosome (CSN) is an evolutionarily conserved eight-subunit (CSN1-8) protein complex that controls protein ubiquitination by deneddylating Cullin-RING E3 ligases (CRLs). The activation and function of CSN hinges on its structural dynamics, which has been challenging to decipher by

Articles

Sulfoxide-containing MS-cleavable cross-linkers capture protein-protein interactions in native cellular environments, aiding PPI identification.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service