Skip to Content
Merck
All Photos(2)

Key Documents

909424

Sigma-Aldrich

Cu(OTs)2

≥95%

Synonym(s):

Copper(II) tosylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€112.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€112.00

About This Item

Empirical Formula (Hill Notation):
C14H14CuO6S2
CAS Number:
Molecular Weight:
405.93
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

€112.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

SMILES string

[Cu+2].[S](=O)(=O)([O-])c2ccc(cc2)C.[S](=O)(=O)([O-])c1ccc(cc1)C

InChI

1S/2C7H8O3S.Cu/c2*1-6-2-4-7(5-3-6)11(8,9)10;/h2*2-5H,1H3,(H,8,9,10);/q;;+2/p-2

InChI key

MRYMYQPDGZIGDM-UHFFFAOYSA-L

Application

Copper(II) tosylate is a copper (II) catalyst used for a variety of transformations, including Mannich condensation, Friedel-Crafts reaction, and Henry reaction.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew G Capacci et al.
Nature chemistry, 9(11), 1073-1077 (2017-10-25)
Although the α-alkylation of ketones has already been established, the analogous reaction using aldehyde substrates has proven surprisingly elusive. Despite the structural similarities between the two classes of compounds, the sensitivity and unique reactivity of the aldehyde functionality has typically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service