Skip to Content
Merck
All Photos(1)

Key Documents

909149

Sigma-Aldrich

Poly(ethylene glycol) diamine

average Mn 400

Synonym(s):

Poly(ethylene glycol) bis(amine), Polyethylene glycol, O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€250.00

€250.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€250.00

About This Item

Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:
MDL number:
UNSPSC Code:
43211915
NACRES:
NA.23

€250.00


Check Cart for Availability

Request a Bulk Order

form

viscous liquid

mol wt

average Mn 400 (by NMR)
average Mn 400

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

color

colorless to pale yellow

Ω-end

amine

α-end

amine

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

NCCOCCOCCN

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

PEGylated materials have found broad use in drug delivery systems, virology, and immunology, as the incorporation of PEG improves pharmacological properties such as increased water solubility, enhanced resistance to degradation (protein hydrolysis), increased circulation half-life, and reduced antigenicity. Polymer for preparing enzyme conjugates soluble in organic solvents; promising drug carrier.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Attaching folic acid on gold nanoparticles using noncovalent interaction via different polyethylene glycol backbones and targeting of cancer cells.
Bhattacharya R, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 3, 224-238 (2007)
Facile Synthesis of a Degradable Poly(ethylene glycol) Platform with Tunable Acid Sensitivity at Physiologically Relevant pH.
Li Z C, et al.
Macromolecules, 51, 6571-6579 (2018)
Rigidity of Two-Component Hydrogels Prepared from Alginate and Poly(ethylene glycol)-Diamines.
Eiselt P, et al.
Macromolecules, 32 (17), 5561-5566 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service