Skip to Content
Merck
All Photos(1)

Key Documents

909084

Sigma-Aldrich

4-Methacryloxy-2-hydroxybenzophenone

≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€267.00

€267.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€267.00

About This Item

Empirical Formula (Hill Notation):
C17H14O4
CAS Number:
Molecular Weight:
282.29
MDL number:
UNSPSC Code:
43211915
NACRES:
NA.23

€267.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

liquid

contains

3000 ppm MEHQ as stabilizer

color

clearoily

storage temp.

2-8°C

SMILES string

O(c1cc(c(cc1)C(=O)c2ccccc2)O)C(=O)C(=C)C

InChI

1S/C17H14O4/c1-11(2)17(20)21-13-8-9-14(15(18)10-13)16(19)12-6-4-3-5-7-12/h3-10,18H,1H2,2H3

InChI key

IMNBHNRXUAJVQE-UHFFFAOYSA-N

Application

4-Methacryloxy-2-hydroxybenzophenone is a UV absorber that can be used in ophthamic and optical applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service