Skip to Content
Merck
All Photos(3)

Key Documents

908886

Sigma-Aldrich

N-Cyano-4-methoxy-picolinimidamide

≥95%

Synonym(s):

Weix ligand

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€141.00
1 G
€394.00

€141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€141.00
1 G
€394.00

About This Item

Empirical Formula (Hill Notation):
C8H8N4O
CAS Number:
Molecular Weight:
176.18
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

€141.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

220-225 °C

SMILES string

N(C(=N)c1nccc(c1)OC)C#N

InChI

1S/C8H8N4O/c1-13-6-2-3-11-7(4-6)8(10)12-5-9/h2-4H,1H3,(H2,10,12)

InChI key

LEWHOCRDELQPCV-UHFFFAOYSA-N

Application

This pyridyl carboxamidine ligand has been demonstrated by Daniel Weix′s lab to enable nickel-catalyzed cross-coupling of diverse basic nitrogen heterocycles with primary and secondary alkyl halides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric C Hansen et al.
Nature chemistry, 8(12), 1126-1130 (2016-11-23)
Ligands are essential for controlling the reactivity and selectivity of reactions catalysed by transition metals. Access to large phosphine ligand libraries has become an essential tool for the application of metal-catalysed reactions industrially, but these existing libraries are not well

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service