Skip to Content
Merck
All Photos(1)

Key Documents

908630

Sigma-Aldrich

Poly(lactide-co-glycolide)-Flamma Fluor near-IR

lactide:glycolide (50:50), Mn 20,000-30,000

Synonym(s):

PLGA near IR, PLGA-FPI749, near IR conjugated PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€261.00

€261.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€261.00

About This Item

Linear Formula:
(C3H4O2)x(C2H2O2)y
UNSPSC Code:
12162002
NACRES:
NA.23

€261.00


Check Cart for Availability

Request a Bulk Order

form

powder or chunks

feed ratio

lactide:glycolide (50:50)

mol wt

Mn 20,000-30,000
average Mn 20,000-30,000

composition

Dye Content, 7.54 μg/mg (polymer)

color

blue to teal

storage temp.

−20°C

Application

Poly(lactide-co-glycolide)-Flamma Fluor near-IR is labelled with Flamma Fluor FPI749 near-IR dye. It can be used in the formation of fluoresent nanoparticles for in-vivo imaging applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanfei Meng et al.
ACS nano, 12(7), 6458-6468 (2018-06-20)
Fluorescence-based whole-body imaging is widely used in the evaluation of nanoparticles (NPs) in small animals, often combined with quantitative analysis to indicate their spatiotemporal distribution following systemic administration. An underlying assumption is that the fluorescence label represents NPs and the
Yihan Xu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1692-1716 (2016-04-22)
Poly (lactic-co-glycolic acid) (PLGA) copolymers have been broadly used in controlled drug release applications. Because these polymers are biodegradable, they provide an attractive option for drug delivery vehicles. There are a variety of material, processing, and physiological factors that impact
R Gref et al.
Science (New York, N.Y.), 263(5153), 1600-1603 (1994-03-18)
Injectable nanoparticulate carriers have important potential applications such as site-specific drug delivery or medical imaging. Conventional carriers, however, cannot generally be used because they are eliminated by the reticulo-endothelial system within seconds or minutes after intravenous injection. To address these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service