Skip to Content
Merck
All Photos(3)

Key Documents

907847

Sigma-Aldrich

HB(trip)2

≥95%

Synonym(s):

Bis(2,4,6-triisopropylphenyl)borane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€589.00

€589.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
€589.00

About This Item

Empirical Formula (Hill Notation):
C30H47B
CAS Number:
Molecular Weight:
418.51
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€589.00


Estimated to ship on16 April 2025


Request a Bulk Order

Assay

≥95%

form

solid

mp

109-110 °C

storage temp.

−20°C

SMILES string

B(c2c(cc(cc2C(C)C)C(C)C)C(C)C)c1c(cc(cc1C(C)C)C(C)C)C(C)C

InChI

1S/C30H47B/c1-17(2)23-13-25(19(5)6)29(26(14-23)20(7)8)31-30-27(21(9)10)15-24(18(3)4)16-28(30)22(11)12/h13-22,31H,1-12H3

InChI key

YGFOJGHLKBDBDV-UHFFFAOYSA-N

Application

HB(trip)2 has been shown by the Hoveyda group to be a useful traceless protecting group for catalytic Z- and E-selective olefin metathesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yucheng Mu et al.
Angewandte Chemie (International ed. in English), 58(16), 5365-5370 (2019-02-13)
An operationally simple in situ protection/deprotection strategy that significantly expands the scope of kinetically controlled catalytic Z- and E-selective olefin metathesis is introduced. Prior to the addition of a sensitive Mo- or Ru-based complex, treatment of a hydroxy- or a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service