Skip to Content
Merck
All Photos(1)

Key Documents

907049

Sigma-Aldrich

Poly(ethylene glycol) dimethacrylate

average MN 4,000 (by NMR), average MN 4,000, cross-linking reagent polymerization reactions, methacrylate, MEHQ as inhibitor, ≤150 ppm MEHQ as inhibitor

Synonym(s):

PEG bismethacrylate, PEG hydrogel, Polyethylene glycol, PEG dimethacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€273.00

€273.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€273.00

About This Item

Linear Formula:
C3H5C(O)(OCH2CH2)nOC(O)C3H5
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€273.00


Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) dimethacrylate, average Mn 4,000, contains MEHQ as inhibitor

form

powder

mol wt

average Mn 4,000 (by NMR)
average Mn 4,000

contains

MEHQ as inhibitor
≤150 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

color

white to off-white

bp

>200 °C/2 mmHg (lit.)

Ω-end

methacrylate

α-end

methacrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

SMILES string

OCCO.CC(=C)C(O)=O

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High purity PEG dimethacrylates are commonly used in PEG-based hydrogels and crosslinking hydrophilic polymers. It is good for tissue engineering and 3D printing applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service