Skip to Content
Merck
All Photos(2)

Key Documents

905283

Sigma-Aldrich

(6-Aminohexyl)triphenylphosphonium bromide hydrobromide

≥95%

Synonym(s):

Aminohexane TPP mitochondrial tag, Mitochondria-targeting probe building block, Phosphonium lipocation tag

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€231.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€231.00

About This Item

Empirical Formula (Hill Notation):
C24H29BrNP · xHBr
Molecular Weight:
442.37 (free base basis)
UNSPSC Code:
12352101
NACRES:
NA.22

€231.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

solid

reaction suitability

reagent type: ligand

functional group

phosphine

storage temp.

2-8°C

Application

Mitochondria perform several functions in the cell, and dysfunction has been implicated in various organ systems and disease states, including neurodegenerative disorders and cancers. The targeting of small molecules to the mitochondria can help to probe and better understand this relationship. This aminohexane triphenylphosphine (TPP) tag is conjugated to small molecules to localize them to the mitochondria in cells where the TPP cations facilitate both cellular uptake and accumulation in the mitochondria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michael P Murphy
Biochimica et biophysica acta, 1777(7-8), 1028-1031 (2008-04-29)
Mitochondrial function and dysfunction contributes to a range of important aspects of biomedical research. Consequently there is considerable interest in developing approaches to modify and report on mitochondria in cells and in vivo. One approach has been to target bioactive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service