Skip to Content
Merck
All Photos(1)

Key Documents

904716

Sigma-Aldrich

(CAAC-Cy)Rh(COD)Cl

Synonym(s):

[2-[2,6-bis(1-methylethyl)phenyl]-3,3-dimethyl-2-azaspiro[4.5]dec-1-ylidene]chloro[(1,2,5,6-η)-1,5-cyclooctadiene]Rhodium

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€350.00

€350.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€350.00

About This Item

Empirical Formula (Hill Notation):
C31H47ClNRh
CAS Number:
Molecular Weight:
572.07
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€350.00


Check Cart for Availability

Request a Bulk Order

Application

(CAAC-Cy)Rh(COD)Cl is a selective hydrogenation catalyst used for the hydrogenation of a variety of arenes including fluoroarenes giving access to all-cis-multifluorinated cycloalkanes.1

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Wei et al.
Journal of the American Chemical Society, 137(29), 9250-9253 (2015-07-15)
Air-stable Rh complexes ligated by strongly σ-donating cyclic (amino)(alkyl)carbenes (CAACs) show unique catalytic activity for the selective hydrogenation of aromatic ketones and phenols by reducing the aryl groups. The use of CAAC ligands is essential for achieving high selectivity and
Mario P Wiesenfeldt et al.
Angewandte Chemie (International ed. in English), 57(27), 8297-8300 (2018-05-24)
We report a method to convert readily available silylated arenes into silylated saturated carbo- and heterocycles by arene hydrogenation. The scope includes alkoxy- and halosilyl substituents. Silyl groups can be derivatized into a plethora of functionalities and find application in
Mario P Wiesenfeldt et al.
Science (New York, N.Y.), 357(6354), 908-912 (2017-08-12)
All-cis-multifluorinated cycloalkanes exhibit intriguing electronic properties. In particular, they display extremely high dipole moments perpendicular to the aliphatic ring, making them highly desired motifs in material science. Very few such motifs have been prepared, as their syntheses require multistep sequences

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service